siRNA Details
| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007378.1 | NP_031404.1 | 2 | 233 | ||
| GenBank mRNA | AF000149.1 | - | 2 | 233 | ||
| AK084860.1 | - | 3 | 461 | |||
| BC057853.1 | - | 2 | 233 | 6492139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca4| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007378.1 | NP_031404.1 | 2 | 233 | ||
| GenBank mRNA | AF000149.1 | - | 2 | 233 | ||
| AK084860.1 | - | 3 | 461 | |||
| BC057853.1 | - | 2 | 233 | 6492139 | ||