siRNA Details
| Gene Symbol: Chrna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007389.5 | NP_031415.2 | 3, 4 | 293 | ||
| GenBank mRNA | AK029177.1 | - | 3, 4 | 262 | ||
| AK132589.1 | - | 3, 4 | 233 | |||
| BC125439.1 | - | 3, 4 | 293 | 40129954 | ||
| BC137678.1 | - | 3, 4 | 293 | 8860697 | ||
| EF520684.1 | - | 2, 3 | 168 | |||
| X03986.1 | - | 3, 4 | 250 | |||

Chrna1
Genomic Map