siRNA Details
| Gene Symbol: UBE2K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111112.1 | NP_001104582.1 | 6 | 2060 | ||
| NM_001111113.1 | NP_001104583.1 | 5 | 2036 | |||
| NM_001312646.1 | NP_001299575.1 | 5 | 2006 | |||
| NM_001312647.1 | NP_001299576.1 | 7 | 2101 | |||
| NM_001312648.1 | NP_001299577.1 | 6 | 2095 | |||
| NM_005339.4 | NP_005330.1 | 7 | 2189 | |||
| GenBank mRNA | AK093533.1 | - | 7 | 2011 | ||
| U58522.1 | - | 7 | 1910 | |||

UBE2K
Genomic Map