siRNA Details
| Gene Symbol: Adam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272057.1 | NP_001258986.1 | 6 | 789 | ||
| NM_001272058.1 | NP_001258987.1 | 6 | 789 | |||
| NM_001272059.1 | NP_001258988.1 | 6 | 789 | |||
| NM_007401.3 | NP_031427.2 | 6 | 789 | |||
| GenBank mRNA | AK016574.1 | - | 6 | 445 | ||
| AK029519.1 | - | 6 | 479 | |||
| AK161262.1 | - | 6 | 789 | |||
| AK161415.1 | - | 3 | 167 | |||
| AK161417.1 | - | 6 | 459 | |||
| BC094237.1 | - | 6 | 486 | 30604517 | ||
| U22059.1 | - | 6 | 453 | |||

Adam5
Genomic Map