siRNA Details
| Gene Symbol: Adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007409.2 | NP_031435.1 | 3 | 232 | ||
| GenBank mRNA | AK146094.1 | - | 3 | 233 | ||
| BC013477.1 | - | 3 | 232 | 4238555 | ||
| BC054467.1 | - | 3 | 212 | 6530906 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adh1| Gene Symbol: Adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007409.2 | NP_031435.1 | 3 | 232 | ||
| GenBank mRNA | AK146094.1 | - | 3 | 233 | ||
| BC013477.1 | - | 3 | 232 | 4238555 | ||
| BC054467.1 | - | 3 | 212 | 6530906 | ||