siRNA Details
| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007423.4 | NP_031449.3 | 1 | 141 | ||
| GenBank mRNA | AK010934.1 | - | 1 | 117 | ||
| AK075972.1 | - | 1 | 121 | |||
| AK076053.1 | - | 1 | 117 | |||
| AK076197.1 | - | 1 | 119 | |||
| AK145975.1 | - | 1 | 117 | |||
| AK146513.1 | - | 1 | 119 | |||
| AK146524.1 | - | 1 | 119 | |||
| AK167549.1 | - | 1 | 117 | |||
| AK168426.1 | - | 1 | 119 | |||
| AK168574.1 | - | 1 | 119 | |||
| AK168576.1 | - | 1 | 117 | |||
| AK168751.1 | - | 1 | 119 | |||
| AK168883.1 | - | 1 | 117 | |||
| AK169046.1 | - | 1 | 119 | |||
| AK169054.1 | - | 1 | 119 | |||
| AK169147.1 | - | 1 | 117 | |||
| BC066206.1 | - | 1 | 104 | 30469872 | ||
| BY773400.1 | - | 1 | 148 | |||
| V00743.1 | - | 1 | 115 | |||

Afp
Genomic Map