siRNA Details
| Gene Symbol: MINOS1-NBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204088.1 | NP_001191017.1 | 5 | 1760 | ||
| NM_001204089.1 | NP_001191018.1 | 4 | 1575 | |||
| GenBank mRNA | AK289456.1 | - | 4 | 1576 | ||
| Gene Symbol: NBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204084.2 | NP_001191013.1 | 4 | 1683 | ||
| NM_001204085.1 | NP_001191014.1 | 4 | 1601 | |||
| NM_001204086.2 | NP_001191015.1 | 4 | 1710 | |||
| NM_001278164.1 | NP_001265093.1 | 4 | 1592 | |||
| NM_001278165.1 | NP_001265094.1 | 4 | 1659 | |||
| NM_001278166.1 | NP_001265095.1 | 4 | 1503 | |||
| NM_005380.7 | NP_005371.2 | 4 | 1716 | |||
| NM_182744.3 | NP_877421.2 | 4 | 1611 | |||
| GenBank mRNA | BC012037.1 | - | 4 | 1515 | 3874204 | |
| D28124.1 | - | 4 | 1484 | |||

MINOS1-NBL1
Genomic Map