siRNA Details
| Gene Symbol: Anapc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008569.2 | NP_032595.2 | 11 | 1554 | ||
| GenBank mRNA | AK052404.1 | - | 11 | 1535 | ||
| AK054305.1 | - | 11 | 1536 | |||
| AK077847.1 | - | 11 | 1555 | |||
| AK090134.1 | - | 11 | 1537 | |||
| AK149762.1 | - | 11 | 1526 | |||
| AK164804.1 | - | 11 | 1540 | |||
| BC139002.1 | - | 11 | 1535 | 8862024 | ||
| BC145492.1 | - | 11 | 1535 | 9054055 | ||
| X80169.1 | - | 11 | 1539 | |||

Anapc1
Genomic Map