siRNA Details
| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008857.3 | NP_032883.2 | 5 | 624 | ||
| GenBank mRNA | AK144979.1 | - | 5 | 624 | ||
| AK147710.1 | - | 5 | 596 | |||
| AK167411.1 | - | 5 | 593 | |||
| AK220517.1 | - | 5 | 648 | |||
| BC021630.1 | - | 5 | 495 | 3980628 | ||
| D28577.1 | - | 5 | 419 | |||

Prkci
Genomic Map