siRNA Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009148.3 | NP_033174.2 | 2 | 220 | ||
| GenBank mRNA | AF022962.1 | - | 2 | 246 | ||
| AK010695.1 | - | 2 | 235 | |||
| AK014499.1 | - | 2 | 243 | |||
| AK036507.1 | - | 2 | 232 | |||
| AK038125.1 | - | 2 | 243 | |||
| AK039983.1 | - | 2 | 243 | |||
| AK086603.1 | - | 2 | 239 | |||
| AK173235.1 | - | 2 | 223 | |||
| BC034644.1 | - | 2 | 220 | 5355276 | ||

Exoc4
Genomic Map