siRNA Details
| Gene Symbol: Exoc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009148.3 | NP_033174.2 | 12 | 1790 | ||
| GenBank mRNA | AF022962.1 | - | 12 | 1816 | ||
| AK028250.1 | - | 10 | 1522 | |||
| AK039983.1 | - | 12 | 1813 | |||
| AK173235.1 | - | 12 | 1793 | |||
| BC034644.1 | - | 12 | 1790 | 5355276 | ||

Exoc4
Genomic Map