siRNA Details
| Gene Symbol: Ssty1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009220.2 | NP_033246.2 | 3 | 663 | ||
| GenBank mRNA | X05260.1 | - | 3 | 663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssty1| Gene Symbol: Ssty1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009220.2 | NP_033246.2 | 3 | 663 | ||
| GenBank mRNA | X05260.1 | - | 3 | 663 | ||