siRNA Details
| Gene Symbol: Pcgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163307.1 | NP_001156779.1 | 7 | 684 | ||
| NM_001163308.1 | NP_001156780.1 | 6 | 546 | |||
| NM_009545.2 | NP_033571.1 | 9 | 951 | |||
| GenBank mRNA | AF483502.1 | - | 5 | 392 | ||
| AF483503.1 | - | 5 | 392 | |||
| AK142837.1 | - | 6 | 459 | |||
| BC016419.1 | - | 7 | 638 | 4458285 | ||
| D90085.1 | - | 9 | 799 | |||

Pcgf2
Genomic Map