siRNA Details
| Gene Symbol: Ikzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040268.1 | - | 6 | 805 | ||
| AK077815.1 | - | 7 | 1370 | |||
| AK142856.1 | - | 5 | 569 | |||
| AK154175.1 | - | 6 | 3996 | |||
| AK220235.1 | - | 7 | 1038 | |||
| BC138789.1 | - | 6 | 603 | 8861811 | ||
| L03547.1 | - | 6 | 588 | |||
| S74708.1 | - | 4 | 649 | |||

Ikzf1
Genomic Map