siRNA Details
| Gene Symbol: Chrnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009602.4 | NP_033732.2 | 2 | 366 | ||
| GenBank mRNA | AF089739.1 | - | 1 | 56 | ||
| AF145286.1 | - | 2 | 305 | |||
| AF299083.1 | - | 2 | 160 | |||
| AF325349.1 | - | 2 | 126 | |||
| AK049722.1 | - | 2 | 339 | |||
| AK051742.1 | - | 2 | 348 | |||
| AK081254.1 | - | 343 | ||||
| AY574267.1 | - | 2 | 140 | |||
| BC065103.1 | - | 2 | 319 | 6838257 | ||

Chrnb2
Genomic Map