siRNA Details
| Gene Symbol: Adam11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110778.1 | NP_001104248.1 | 15 | 1462 | ||
| NM_009613.2 | NP_033743.2 | 15 | 1462 | |||
| GenBank mRNA | AB009676.1 | - | 15 | 1295 | ||
| AK157926.1 | - | 15 | 1467 | |||
| BC054536.1 | - | 15 | 1322 | 6400084 | ||

Adam11
Genomic Map