siRNA Details
| Gene Symbol: Adam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009620.1 | NP_033750.1 | 1 | 447 | ||
| GenBank mRNA | AK076647.1 | - | 1 | 507 | ||
| AY158688.1 | - | 1 | 447 | |||
| BC150159.1 | - | 1 | 451 | 40141514 | ||
| BC150481.1 | - | 1 | 451 | 40141515 | ||
| Gene Symbol: Gm4787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 518 | ||
| GenBank mRNA | BC141094.1 | - | 1 | 496 | 9055706 | |

Adam4
Genomic Map