siRNA Details
| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009624.3 | NP_033754.2 | 2 | 1272 | ||
| GenBank mRNA | U30602.1 | - | 2 | 579 | ||
| Z50190.1 | - | 1 | 548 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adcy9| Gene Symbol: Adcy9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009624.3 | NP_033754.2 | 2 | 1272 | ||
| GenBank mRNA | U30602.1 | - | 2 | 579 | ||
| Z50190.1 | - | 1 | 548 | |||