siRNA Details
| Gene Symbol: Agtrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301281.1 | NP_001288210.1 | 5 | 1760 | ||
| NM_009642.4 | NP_033772.2 | 5 | 1807 | |||
| GenBank mRNA | AK036598.1 | - | 5 | 1736 | ||
| AK051014.1 | - | 1 | 225 | |||
| AK152467.1 | - | 5 | 1807 | |||
| BC046820.1 | - | 5 | 1793 | 6400135 | ||

Agtrap
Genomic Map