siRNA Details
| Gene Symbol: Akap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009650.2 | NP_033780.2 | 5 | 1229 | ||
| GenBank mRNA | AF093406.1 | - | 5 | 1155 | ||
| BC100458.1 | - | 5 | 1102 | 30917596 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap3| Gene Symbol: Akap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009650.2 | NP_033780.2 | 5 | 1229 | ||
| GenBank mRNA | AF093406.1 | - | 5 | 1155 | ||
| BC100458.1 | - | 5 | 1102 | 30917596 | ||