siRNA Details
| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010657.3 | NP_034787.2 | 4 | 1953 | ||
| GenBank mRNA | AY454345.1 | - | 5 | 1900 | ||
| BC145272.1 | - | 1 | 1472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep3| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010657.3 | NP_034787.2 | 4 | 1953 | ||
| GenBank mRNA | AY454345.1 | - | 5 | 1900 | ||
| BC145272.1 | - | 1 | 1472 | |||