siRNA Details
| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011150.2 | NP_035280.1 | 6 | 1171 | ||
| GenBank mRNA | AK079915.1 | - | 6 | 1200 | ||
| AK137836.1 | - | 6 | 1148 | |||
| AK150021.1 | - | 6 | 1200 | |||
| AK152182.1 | - | 6 | 1113 | |||
| AK153313.1 | - | 6 | 1187 | |||
| AK153396.1 | - | 6 | 1189 | |||
| AK155031.1 | - | 6 | 1113 | |||
| BC090658.1 | - | 6 | 1167 | 30666981 | ||
| L16894.1 | - | 6 | 1178 | |||
| X67809.1 | - | 6 | 1171 | |||

Lgals3bp
Genomic Map