siRNA Details
| Gene Symbol: Ikzf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011770.4 | NP_035900.2 | 8 | 1667 | ||
| GenBank mRNA | AF044257.1 | - | 8 | 1562 | ||
| AK047925.1 | - | 8 | 1529 | |||
| BC138606.1 | - | 8 | 1507 | 8861626 | ||
| BC138608.1 | - | 8 | 1507 | 8861628 | ||

Ikzf2
Genomic Map