siRNA Details
| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013454.3 | NP_038482.3 | 44, 45 | 6255 | ||
| GenBank mRNA | CF745062.1 | - | 6, 7 | 656 | ||
| CN702062.1 | - | 2, 3 | 144 | |||
| X75926.1 | - | 43, 44 | 6018 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca1| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013454.3 | NP_038482.3 | 44, 45 | 6255 | ||
| GenBank mRNA | CF745062.1 | - | 6, 7 | 656 | ||
| CN702062.1 | - | 2, 3 | 144 | |||
| X75926.1 | - | 43, 44 | 6018 | |||