siRNA Details
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013456.2 | NP_038484.1 | 7 | 702 | ||
GenBank mRNA | AF093775.1 | - | 7 | 677 | ||
BC111890.1 | - | 7 | 668 | 40060598 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Actn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013456.2 | NP_038484.1 | 7 | 702 | ||
GenBank mRNA | AF093775.1 | - | 7 | 677 | ||
BC111890.1 | - | 7 | 668 | 40060598 |