siRNA Details
| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013456.2 | NP_038484.1 | 10, 11 | 1179 | ||
| GenBank mRNA | AF093775.1 | - | 10, 11 | 1154 | ||
| BC111890.1 | - | 10, 11 | 1145 | 40060598 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actn3| Gene Symbol: Actn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013456.2 | NP_038484.1 | 10, 11 | 1179 | ||
| GenBank mRNA | AF093775.1 | - | 10, 11 | 1154 | ||
| BC111890.1 | - | 10, 11 | 1145 | 40060598 | ||