siRNA Details
| Gene Symbol: Gdf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013526.1 | NP_038554.1 | 2 | 3458 | ||
| GenBank mRNA | AJ537425.1 | - | 2 | 3458 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf6| Gene Symbol: Gdf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013526.1 | NP_038554.1 | 2 | 3458 | ||
| GenBank mRNA | AJ537425.1 | - | 2 | 3458 | ||