siRNA Details
| Gene Symbol: Greb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252071.1 | NP_001239000.1 | 2, 3 | 457 | ||
| NM_015764.4 | NP_056579.2 | 4, 5 | 779 | |||
| GenBank mRNA | AF180470.1 | - | 3, 4 | 435 | ||
| AK045057.1 | - | 1, 2 | 233 | |||
| AK147337.1 | - | 2, 3 | 307 | |||
| AK157808.1 | - | 2, 3 | 391 | |||
| BC160931.1 | - | 3, 4 | 389 | |||

Greb1
Genomic Map