siRNA Details
| Gene Symbol: Il27ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016671.3 | NP_057880.2 | 11, 12 | 1552 | ||
| GenBank mRNA | AF053005.1 | - | 11, 12 | 1520 | ||
| AF265243.1 | - | 11, 12 | 1531 | |||
| AK040457.1 | - | 9, 10 | 1892 | |||
| AK133753.1 | - | 11, 12 | 1567 | |||
| BC032878.1 | - | 11, 12 | 1552 | 1244998 | ||

Il27ra
Genomic Map