siRNA Details
| Gene Symbol: Sult1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016771.3 | NP_058051.3 | 5 | 615 | ||
| GenBank mRNA | AF026073.1 | - | 5 | 493 | ||
| AK133530.1 | - | 5 | 534 | |||
| AK160466.1 | - | 5 | 600 | |||
| BC010752.1 | - | 5 | 588 | |||
| BC066190.1 | - | 5 | 568 | 30472198 | ||
| U32371.1 | - | 5 | 565 | |||

Sult1d1
Genomic Map