siRNA Details
| Gene Symbol: WDR46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164267.1 | NP_001157739.1 | 4 | 617 | ||
| NM_005452.5 | NP_005443.3 | 4 | 779 | |||
| XM_017011484.1 | XP_016866973.1 | 4 | 493 | |||
| XM_017011485.1 | XP_016866974.1 | 2 | 141 | |||
| XM_017011487.1 | XP_016866976.1 | 2 | 141 | |||
| GenBank mRNA | AF447870.1 | - | 4 | 471 | ||
| AK292171.1 | - | 4 | 762 | |||
| AK297925.1 | - | 4 | 374 | |||
| AK298151.1 | - | 4 | 328 | |||
| AK308534.1 | - | 3 | 255 | |||
| AK315616.1 | - | 4 | 491 | |||
| BC000388.2 | - | 4 | 466 | 2821198 | ||
| BX439421.2 | - | 4 | 466 | |||
| DB272479.1 | - | 4 | 490 | |||

WDR46
Genomic Map