siRNA Details
| Gene Symbol: Ostf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017375.3 | NP_059071.1 | 10 | 993 | ||
| GenBank mRNA | AK002899.1 | - | 10 | 972 | ||
| AK005525.1 | - | 10 | 962 | |||
| AK008123.1 | - | 10 | 941 | |||
| AK008467.1 | - | 10 | 941 | |||
| AK010074.1 | - | 10 | 989 | |||
| AK145949.1 | - | 10 | 944 | |||
| AK149175.1 | - | 971 | ||||
| AK150145.1 | - | 10 | 963 | |||
| AK155028.1 | - | 10 | 963 | |||
| AK205236.1 | - | 1 | 50 | |||
| AK207431.1 | - | 1 | 131 | |||
| AK209921.1 | - | 1 | 131 | |||
| AK210606.1 | - | 1 | 131 | |||
| AK212936.1 | - | 1 | 131 | |||
| BC060986.1 | - | 10 | 872 | 30300544 | ||
| U58888.1 | - | 10 | 941 | |||

Ostf1
Genomic Map