siRNA Details
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 4 | 253 | ||
GenBank mRNA | AB028921.1 | - | 4 | 246 | ||
AK011325.1 | - | 4 | 258 | |||
AK203044.1 | - | 3 | 126 | |||
BC010401.1 | - | 4 | 303 | |||
BC046801.1 | - | 4 | 242 | 6403966 | ||
BC051015.1 | - | 4 | 253 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013946, XM_006241055, BC085785.
Panther Classification:
Gene Ontology Categories:
