siRNA Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017476.2 | NP_059504.2 | 5 | 517 | ||
| GenBank mRNA | AB028921.1 | - | 5 | 510 | ||
| AK011325.1 | - | 5 | 522 | |||
| AK166526.1 | - | 6 | 433 | |||
| AK195897.1 | - | 2 | 274 | |||
| BC046801.1 | - | 5 | 506 | 6403966 | ||
| BC051015.1 | - | 5 | 517 | |||

Akap8l
Genomic Map