siRNA Details
| Gene Symbol: Sertad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018820.4 | NP_061290.1 | 2 | 858 | ||
| GenBank mRNA | AF218291.1 | - | 2 | 765 | ||
| AF366401.1 | - | 2 | 901 | |||
| AK002324.1 | - | 2 | 859 | |||
| AK004022.1 | - | 2 | 858 | |||
| BC016077.1 | - | 2 | 818 | 4486318 | ||

Sertad1
Genomic Map