siRNA Details
| Gene Symbol: Arid3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019689.2 | NP_062663.1 | 7 | 1584 | ||
| GenBank mRNA | AF116847.1 | - | 7 | 1584 | ||
| AK084952.1 | - | 5 | 905 | |||
| AK139455.1 | - | 8 | 2838 | |||
| AK148073.1 | - | 8 | 1682 | |||
| BC145330.1 | - | 7 | 1648 | |||
| BC150755.1 | - | 7 | 1578 | 9087666 | ||

Arid3b
Genomic Map