siRNA Details
| Gene Symbol: Tusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019742.4 | NP_062716.1 | 3 | 689 | ||
| GenBank mRNA | AF123387.1 | - | 3 | 576 | ||
| AK082684.1 | - | 3 | 690 | |||
| AK198946.1 | - | 1 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tusc2| Gene Symbol: Tusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019742.4 | NP_062716.1 | 3 | 689 | ||
| GenBank mRNA | AF123387.1 | - | 3 | 576 | ||
| AK082684.1 | - | 3 | 690 | |||
| AK198946.1 | - | 1 | 265 | |||