siRNA Details
| Gene Symbol: Higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019814.4 | NP_062788.1 | 4 | 909 | ||
| GenBank mRNA | AB253941.1 | - | 3 | 421 | ||
| AK008756.1 | - | 4 | 536 | |||
| AK018360.1 | - | 4 | 537 | |||
| AY028386.1 | - | 4 | 462 | |||
| AY028387.1 | - | 3 | 946 | |||
| BC132371.1 | - | 4 | 527 | 40130648 | ||
| BC151191.1 | - | 4 | 527 | 8861324 | ||

Higd1a
Genomic Map