siRNA Details
| Gene Symbol: Gltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019821.2 | NP_062795.2 | 5 | 694 | ||
| GenBank mRNA | AK003218.1 | - | 1 | 161 | ||
| AK157313.1 | - | 5 | 681 | |||
| BC016584.1 | - | 5 | 694 | 3157800 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gltp| Gene Symbol: Gltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019821.2 | NP_062795.2 | 5 | 694 | ||
| GenBank mRNA | AK003218.1 | - | 1 | 161 | ||
| AK157313.1 | - | 5 | 681 | |||
| BC016584.1 | - | 5 | 694 | 3157800 | ||