siRNA Details
| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019832.3 | NP_062806.1 | 9 | 1027 | ||
| GenBank mRNA | AB033130.1 | - | 8 | 886 | ||
| AK016588.1 | - | 8 | 799 | |||
| BC052347.1 | - | 9 | 1027 | 6774715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gkap1| Gene Symbol: Gkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019832.3 | NP_062806.1 | 9 | 1027 | ||
| GenBank mRNA | AB033130.1 | - | 8 | 886 | ||
| AK016588.1 | - | 8 | 799 | |||
| BC052347.1 | - | 9 | 1027 | 6774715 | ||