siRNA Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021539.4 | NP_067514.2 | 9 | 1285 | ||
| GenBank mRNA | AF033188.1 | - | 8 | 1129 | ||
| AF072881.1 | - | 9 | 1206 | |||
| AK136416.1 | - | 8 | 1185 | |||
| AK150710.1 | - | 7 | 859 | |||
| AK157999.1 | - | 9 | 1244 | |||
| BC055100.1 | - | 9 | 1285 | 6589250 | ||

Wsb2
Genomic Map