siRNA Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021713.2 | NP_068359.1 | 1, 2 | 256 | ||
| GenBank mRNA | AF289484.1 | - | 1, 2 | 248 | ||
| AK002656.1 | - | 1, 2 | 231 | |||
| AK004104.1 | - | 1, 2 | 231 | |||
| AK013232.1 | - | 1, 2 | 202 | |||
| AK017955.1 | - | 1 | 370 | |||
| BC023124.1 | - | 1 | 363 | |||
| BC028501.1 | - | 1, 2 | 241 | 5256915 | ||
| CJ180315.1 | - | 1, 2 | 256 | |||

Myg1
Genomic Map