siRNA Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023168.3 | NP_075657.1 | 7 | 1066 | ||
| GenBank mRNA | AF182040.1 | - | 7 | 1049 | ||
| AK002347.1 | - | 3 | 242 | |||
| AK003920.1 | - | 7 | 1070 | |||
| AK186528.1 | - | 2 | 202 | |||
| AK218437.1 | - | 4 | 367 | |||
| BC010802.1 | - | 7 | 1099 | 4236429 | ||
| BC019157.1 | - | 7 | 1087 | |||
| BC037667.1 | - | 7 | 1066 | 5376054 | ||

Grina
Genomic Map