siRNA Details
| Gene Symbol: Rpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042556.1 | NP_001036021.1 | 3 | 594 | ||
| NM_023323.3 | NP_075812.3 | 3 | 244 | |||
| GenBank mRNA | AB041810.1 | - | 3 | 624 | ||
| AK010776.1 | - | 3 | 238 | |||
| AK011644.1 | - | 3 | 222 | |||
| AK012188.1 | - | 3 | 244 | |||
| AK012641.1 | - | 3 | 223 | |||
| AK013395.1 | - | 3 | 219 | |||
| AK017678.1 | - | 3 | 215 | |||
| AK020030.1 | - | 3 | 205 | |||
| AK051135.1 | - | 3 | 595 | |||
| BQ828311.1 | - | 3 | 255 | |||

Rpf2
Genomic Map