siRNA Details
| Gene Symbol: Pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023530.2 | NP_076019.2 | 3, 4 | 560 | ||
| GenBank mRNA | AF339738.1 | - | 3, 4 | 582 | ||
| AK008035.1 | - | 3, 4 | 586 | |||
| AY358033.1 | - | 3, 4 | 589 | |||
| BC021592.1 | - | 3, 4 | 560 | 5133141 | ||

Pla2g12b
Genomic Map