siRNA Details
| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023729.3 | NP_076218.3 | 13 | 1700 | ||
| GenBank mRNA | AF459789.1 | - | 13 | 1694 | ||
| AK016595.1 | - | 13 | 1694 | |||
| BB020038.1 | - | 1 | 204 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asz1| Gene Symbol: Asz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023729.3 | NP_076218.3 | 13 | 1700 | ||
| GenBank mRNA | AF459789.1 | - | 13 | 1694 | ||
| AK016595.1 | - | 13 | 1694 | |||
| BB020038.1 | - | 1 | 204 | |||