siRNA Details
| Gene Symbol: Uba7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023738.4 | NP_076227.1 | 5 | 618 | ||
| GenBank mRNA | AK004894.2 | - | 5 | 619 | ||
| AK054410.1 | - | 5 | 625 | |||
| AK089235.1 | - | 5 | 622 | |||
| AK129009.1 | - | 5 | 530 | |||
| AK170421.1 | - | 5 | 611 | |||
| AK170644.1 | - | 5 | 617 | |||
| AK172467.1 | - | 5 | 580 | |||
| BC037693.1 | - | 3 | 392 | |||
| BC138843.1 | - | 5 | 535 | 8861865 | ||

Uba7
Genomic Map