siRNA Details
| Gene Symbol: Cuedc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164290.1 | NP_001157762.1 | 10 | 938 | ||
| NM_001164291.1 | NP_001157763.1 | 9 | 830 | |||
| NM_001164292.1 | NP_001157764.1 | 9 | 827 | |||
| NM_001164293.1 | NP_001157765.1 | 9 | 797 | |||
| NM_001164294.1 | NP_001157766.1 | 9 | 776 | |||
| NM_001164295.1 | NP_001157767.1 | 9 | 773 | |||
| NM_024192.2 | NP_077154.2 | 10 | 929 | |||
| GenBank mRNA | AB232359.1 | - | 8 | 728 | ||
| AK003457.1 | - | 9 | 732 | |||
| AK013884.1 | - | 9 | 788 | |||
| BC003442.1 | - | 10 | 893 | 2650719 | ||

Cuedc2
Genomic Map