siRNA Details
| Gene Symbol: Sec13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024206.4 | NP_077168.2 | 5, 6 | 491 | ||
| GenBank mRNA | AK003354.1 | - | 5, 6 | 492 | ||
| AK009755.1 | - | 4, 5 | 475 | |||
| BC002128.1 | - | 5, 6 | 494 | 3590767 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec13| Gene Symbol: Sec13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024206.4 | NP_077168.2 | 5, 6 | 491 | ||
| GenBank mRNA | AK003354.1 | - | 5, 6 | 492 | ||
| AK009755.1 | - | 4, 5 | 475 | |||
| BC002128.1 | - | 5, 6 | 494 | 3590767 | ||