siRNA Details
| Gene Symbol: Anapc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024213.2 | NP_077175.1 | 9, 10 | 915 | ||
| GenBank mRNA | AK088759.1 | - | 9, 10 | 905 | ||
| AK146000.1 | - | 9, 10 | 912 | |||
| AK148625.1 | - | 9, 10 | 925 | |||
| AK166061.1 | - | 9, 10 | 1038 | |||
| AK168031.1 | - | 9, 10 | 912 | |||
| AK168897.1 | - | 10, 11 | 997 | |||
| AK192250.1 | - | 4, 5 | 307 | |||
| BC016237.1 | - | 9, 10 | 898 | 4504779 | ||
| BC024870.1 | - | 9, 10 | 915 | 4949079 | ||

Anapc4
Genomic Map